Orders LinkContactUsLinkComponent
Steriliseringsposer
Steriliseringsposer/poser til bortskaffelse, autoklaverbare (121 °C), PP, 300×200 mm
  129-0131
 :   Brand
Steriliseringsposer
  129-0131
 :  Brand
Steriliseringsposer
Steriliseringsposer
Steriliseringsposer/poser til bortskaffelse, autoklaverbare (121 °C), PP, 300×200 mm
198,00 kr
1 * 100 Items
Bags: Disposal bag, PP/PA.
Holders:
Poseholdere fremstillet af stålwire belagt med epoxyharpiks. Gummifod. Ikke autoklaverbar.
Epoxy-coated steel wire, with rubber feet. Non-autoclavable.
 : risk of injury. Never put sharp objects such as needles into plastic bags. Bags should stay open during sterilisation.
  • Beskrivelse:
    Steriliseringsposer/poser til bortskaffelse, autoklaverbare (121 °C)
  • L×B:
    300×200 mm
  • Materiale:
    PP
  • Seq:
    01