Kolophonium
Glass bottle for solids, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: Terpentinoliefri harpiks
- CAS nummer:8050-09-7
- MDL nummer:MFCD00132205
- Smeltende Pt:75…81 °C
- Tæthed:1,1 g/cm³ (20 °C)
- Flash Pt:180 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:232-475-7
Specifikationer
Specifikationstestresultater
- Appearance:Almost white to yellow to orange Powder or flakes or chunks
- Infrared spectrum:Conforms
- Melting point:72 to 83 °C
- Acid value:156 to 182 mg KOH/g
- Uniformity coefficient:≤1.8
- Moisture holding capacity:53 to 58 % (H+)
- Total volume capacity:≥1.8 eq/l (H+)
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:500 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



