Pyronine G, certified
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
Pyronine G, certified
Katalog # ACRO418630050
Leverandør: Thermo Fisher Scientific
CAS nummer: 92-32-0
Gå til:
Product Details & Documents
Synonymer: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
- CAS nummer:92-32-0
- Formel:C₁₇H₁₉ClN₂O
- MDL nummer:MFCD00011725
- Molecular Weight:302.8 g/mol
- Merck Index:12,08190
- Smeltende Pt:250…260 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-147-8
Documents
ACRO41863
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥45 %
- UV Lambda max:546 to 551 nm
- Absorption Ratio P-15/P+15:1.23 to 1.7
- BSC Certification:Passes
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g



