Pyronine G, certified
Glass bottle for solids, 1 g
Leverandør: Thermo Fisher Scientific
Pyronine G, certified
Katalog # ACRO418630010
Leverandør: Thermo Fisher Scientific
CAS nummer: 92-32-0
Gå til:
Product Details & Documents
Synonymer: Pyronin Y, Pyronine Y, 3,6-Bis(dimethylamino)xanthylium chloride
- CAS nummer:92-32-0
- Formel:C₁₇H₁₉ClN₂O
- MDL nummer:MFCD00011725
- Molecular Weight:302.8 g/mol
- Merck Index:12,08190
- Smeltende Pt:250…260 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-147-8
Documents
ACRO41863
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Dark green Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥45 %
- UV Lambda max:546 to 551 nm
- Absorption Ratio P-15/P+15:1.23 to 1.7
- BSC Certification:Passes
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:1 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



