Orders LinkContactUsLinkComponent
Accessories for pipette tips, PD-Tips
Accessories for pipette tips, PD-Tips
Adapters, PP, autoclavable, non sterile, 25 and 50 ml PD-Tips and PD-Tips II
Katalog # 612-5809
Leverandør:   Brand
Accessories for pipette tips, PD-Tips
Katalog # 612-5809
Leverandør:  Brand
Accessories for pipette tips, PD-Tips
Accessories for pipette tips, PD-Tips
Adapters, PP, autoclavable, non sterile, 25 and 50 ml PD-Tips and PD-Tips II
Price
214,00 kr
Log ind for tilgængelighed
1 * 10 Items
Specifikationer
  • Til:
    25 and 50 ml PD-Tips and PD-Tips II
  • Beskrivelse:
    Adapters, PP, autoclavable, non sterile