Orders LinkContactUsLinkComponent
Repair set
Repair set
Repair set for Transferpettor™ 100, 200 µl
Katalog # 612-5385
Leverandør:   Brand
Repair set
Katalog # 612-5385
Leverandør:  Brand
Repair set
Repair set
Repair set for Transferpettor™ 100, 200 µl
Price
336,00 kr
Log ind for tilgængelighed
1 * 1 Each
Product Details & Documents
Set includes 1 allen key, 1 piston rod with fitted PTFE Transferpettor™ seal (for capacities ≥ 20 µl), 1 calibration gauge, 1 screwdriver, 3 clamping discs, 1 fixing screw, 3 PTFE Transferpettor™ seals, 1 mounting block (for capacities ≥ 20 µl).
Documents
Specifikationer
  • Beskrivelse:
    Repair set for Transferpettor™ 100, 200 µl
  • Seq:
    05