Phenolphthalein ACS
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- CAS nummer:77-09-8
- Formel:C₂₀H₁₄O₄
- MDL nummer:MFCD00005913
- Molecular Weight:318.33 g/mol
- Merck Index:14,07243
- Smeltende Pt:258…263 °C
- Tæthed:1,3 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-004-7
- NÅ:01-2119498295-24
Specifikationer
Specifikationstestresultater
- Appearance:Off-white Crystalline powder
- Infrared spectrum:Conforms
- Visual transition interval:From pH 8.0 (colorless) to pH 10 (red)
- Clarity of alcohol solution:Passes test
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g



