Phenolphthalein ACS
Plastic bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: 3,3-Bis(4-hydroxyphenyl)isobenzofuran-1(3H)-one
- CAS nummer:77-09-8
- Formel:C₂₀H₁₄O₄
- MDL nummer:MFCD00005913
- Molecular Weight:318.33 g/mol
- Merck Index:14,07243
- Smeltende Pt:258…263 °C
- Tæthed:1,3 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:201-004-7
- NÅ:01-2119498295-24
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Off-white Crystalline powder
- Infrared spectrum:Conforms
- Visual transition interval:From pH 8.0 (colorless) to pH 10 (red)
- Clarity of alcohol solution:Passes test
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



