Ekssikatorplader
PP, 140 mm
BRND66015
: Brand
Desiccator plates for glass and plastic desiccators.
- Central hole measures approximately 20 mm diameter
- Small perforations measure approximately 5 mm diameter
- Ø:140 mm
- Model:PP
- Seq:04
Desiccator plates for glass and plastic desiccators.