Nitrobenzen 99+% ACS
Glass bottle, 25 mL
Leverandør: Thermo Fisher Scientific
Nitrobenzen 99+% ACS
Katalog # ACRO415780250
Leverandør: Thermo Fisher Scientific
CAS nummer: 98-95-3
Gå til:
Product Details & Documents
- CAS nummer:98-95-3
- Formel:C₆H₅NO₂
- MDL nummer:MFCD00007043
- Molecular Weight:123.11 g/mol
- Merck Index:13,06621
- Kogende Pt:210,8 °C (1013 hPa)
- Smeltende Pt:5…6 °C
- Tæthed:1,205 g/cm³
- Flash Pt:88 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:202-716-0
- FN:1662
- ADR:6.1,II
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.5500 to 1.5540 (20°C, 589 nm)
- GC:≥99.0 %
- Residue after evaporation:≤0.005 %
- Chloride (Cl):≤5 ppm
- Water soluble titr. acid:≤0.0005 meq/g
Specifikationer
- Pakning:Glass bottle
- Pakn:25 mL



