Orders LinkContactUsLinkComponent
Farvekar
Farvekar
Farvekar, transparent, 83×101×70 mm
Katalog # BRND474300
Leverandør:   Brand
Farvekar
Katalog # BRND474300
Leverandør:  Brand
Farvekar
Farvekar
Farvekar, transparent, 83×101×70 mm
Price
1.540,00 kr
Log ind for tilgængelighed
1 * 4 Items
Product Details & Documents
PMP, transparent.
Trough comes with two lids: one to help protect against evaporation when not in use; one with a handle slot facilitating the staining process.
PP tray with fold down handle can accomodate 20 slides. The close fitting lid helps protect against evaporation.
Ordering information: Trough and tray (insert) have to be ordered separately.
Specifikationer
  • Beskrivelse:
    Farvekar, transparent
  • B×D×H:
    83×101×70 mm