Hydroxylammoniumchlorid ACS
Plastic bottle, 500 g
Leverandør: Thermo Fisher Scientific
Hydroxylammoniumchlorid ACS
Katalog # ACRO412055000
Leverandør: Thermo Fisher Scientific
CAS nummer: 5470-11-1
Gå til:
Product Details & Documents
Synonymer: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS nummer:5470-11-1
- Formel:NH₂OH·HCl
- MDL nummer:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:13,04853
- Kogende Pt:306 °C (1013 hPa)
- Smeltende Pt:155…158 °C
- Tæthed:1,67 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:226-798-2
- FN:2923
- ADR:8,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Residue after ignition:≤0.05 %
- Titration with KMnO4:≥96.0 %
- Clarity of alcohol solution:Passes test
- Ammonium (NH4):Passes test
- Iron (Fe):≤5 ppm
- Heavy metals (ICP-OES):≤5 ppm
- Sulfur compounds (as SO4):≤0.005 %
- Titrable free acid:≤0.25 meq/g
Specifikationer
- Pakning:Plastic bottle
- Pakn:500 g