Geraniol 99%
Glass bottle, 100 g
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
Synonymer: (E)-3,7-Dimethylocta-2,6-dien-1-ol
- CAS nummer:106-24-1
- Formel:C₁₀H₁₈O
- MDL nummer:MFCD00002917
- Molecular Weight:154.25 g/mol
- Kogende Pt:229…230 °C (757 mmHg)
- Smeltende Pt:–15 °C
- Tæthed:0,878 g/cm³
- Flash Pt:76 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-377-1
Documents
Specifikationer
Specifikationstestresultater
- Appearance:Clear colorless to light yellow Liquid
- Infrared spectrum:Conforms
- Refractive index:1.4750 to 1.4780 (20°C, 589 nm)
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 g



