Centrifugerør, ASTM, BLAUBRAND®
Tidligere standard ASTM D 96, 100 ml, uden cap, 700×g, Cylindrical
Katalog # 525-1271
Leverandør: Brand
Gå til:
Product Details & Documents
Borosilikat 3,3 glas, gradueret.
- Withstands forces up to 700×g
- Design, accuracy etc., to meet ASTM specifications
- For water and sediments in crude oils according to ASTM D91/D96
Specifikationer
- Lukkestørrelse:uden cap
- Model:Cylindrical
- Kapacitet:100 ml
- Beskrivelse:Tidligere standard ASTM D 96
- RCF maks.:700×g