Eosin B, pure, certified
Glass bottle for solids, 25 g
Leverandør: Thermo Fisher Scientific
Eosin B, pure, certified
Katalog # ACRO409420250
Leverandør: Thermo Fisher Scientific
CAS nummer: 548-24-3
Gå til:
Product Details & Documents
Synonymer: Eosin bluish, 2,7-Dinitro-4,5-dibromofluorescein, Eosin B disodium salt, Sodium 2-(4,5-dibromo-2,7-dinitro-6-oxido-3-oxo-3H-xanthen-9-yl)benzoate, 2-(4,5-dibromo-3,6-dihydroxy-2,7-dinitroxanthen-9-yl)-benzoic acid disodium salt, Acid Red 91
- CAS nummer:548-24-3
- Formel:C₂₀H₆Br₂N₂Na₂O₉
- MDL nummer:MFCD00005041
- Molecular Weight:624.06 g/mol
- Merck Index:12,03638
- Smeltende Pt:>310 °C (dec.)
- Opbevaringstemperatur:Stue temperatur
- EINECS:208-943-1
Documents
ACRO40942
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Dark brown to dark red-purple Crystalline powder
- Infrared spectrum:Conforms
- Dye content:≥85 %
- UV Lambda max:511 to 520 nm (in water + 1% Na2CO3)
- Absorption Ratio P-15/P+15:0.96 to 1.22
- BSC Certification:Passes
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



