Orders LinkContactUsLinkComponent
Weighing boats
Weighing boats
25 ml, 70×45×12 mm, hvid
Katalog # BRND155546
Leverandør:   Brand
Weighing boats
Katalog # BRND155546
Leverandør:  Brand
Weighing boats
Weighing boats
25 ml, 70×45×12 mm, hvid
Price
577,00 kr
Log ind for tilgængelighed
1 * 500 Items
Product Details & Documents
Polystyrene, impact resistant, antistatic.
  • Light and flat design
  • Wall thickness approximately 0,2 mm
Documents
Specifikationer
  • Kapacitet:
    25 ml
  • Farve:
    hvid
  • B×D×H:
    70×45×12 mm