Weighing boats
250 ml, 140×140×21 mm, hvid
Katalog # BRND155544
Leverandør: Brand
Gå til:
Product Details & Documents
Polystyrene, impact resistant, antistatic.
- Light and flat design
- Wall thickness approximately 0,2 mm
Documents
BRND155544_TDS_EN
Datablad
Specifikationer
- Kapacitet:250 ml
- Farve:hvid
- B×D×H:140×140×21 mm