Tragte
82 mm, 11 mm, 100 mm
Katalog # BRND148235
Leverandør: Brand
Gå til:
Product Details & Documents
PP, translucent.
- External flutings allow air to escape during filtration
- Excellent chemical resistance, autoclavable
- Suitable for use with standard filter papers
For rapid filtration.
Specifikationer
- Tragt udv. Ø:100 mm
- Stilk udv. Ø:11 mm
- Stilklængde:82 mm