Natriumcholat 99%
Glass bottle for solids, 100 g
: 3α,7α,12α-Trihydroxy-5β-cholanic acid sodium salt, 3α,7α,12α-Trihydroxycholan-24-oic acid sodium salt, 3α,7α,12α-Trihydroxycholanic acid sodium salt, Sodium 3α,7α,12α-trihydroxy-5β-cholanoate, (R)-4-((3R,5S,7R,8R,9S,10S,12S,13R,14S,17R)-3,7,12-Trihydroxy-10,13-dimethylhexadecahydro-1H-cyclopenta[a]phenanthren-17-yl)pentanoic acid sodium salt, 3α,7α,12α-Trihydroxy-5β-cholan-24-oic acid sodium salt
- :361-09-1
- :C₂₄H₃₉NaO₅
- :MFCD00150749
- :430.56 g/mol
- :198 °C
- :Stue temperatur
- :206-643-5
- Appearance:White to off-white Powder
- Infrared spectrum:Conforms
- Loss on drying:≤5.0 % (110°C)
- pH:7.5 to 9.5 (5 % in water)
- Specific optical rotation:+33° to +37° (20°C, 589 nm) (c=0.6, ethanol)
- Titration with HClO4:98.0 to 102.0 % (On dried basis)
- Heavy metals (as Pb):≤50 ppm
- Impurity:≤2.0 % Sodium deoxycholate
- Pakning:Glass bottle for solids
- Pakn:100 g
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



