Orders LinkContactUsLinkComponent
Dropping pipettes
5 ml, 194 mm, 100/pakke
Katalog # BRND125420
Leverandør:   Brand
Dropping pipettes
Katalog # BRND125420
Leverandør:  Brand
Dropping pipettes
Dropping pipettes
5 ml, 194 mm, 100/pakke
Price
296,00 kr
Log ind for tilgængelighed
1 * 100 Items
Product Details & Documents
LDPE, graduated. For sampling and decanting infectious or toxic liquids.
Specifikationer
  • Pakket:
    100/pakke
  • Kapacitet:
    5 ml
  • Længde:
    194 mm