Acetone 99.6% ACS
Glass bottle, 500 mL
Acetone 99.6% ACS
ACRO400105000
: 67-64-1
: Propanon
- :67-64-1
- :CH₃COCH₃
- :MFCD00008765
- :58.08 g/mol
- :13,00067
- :56,2 °C (1013 hPa)
- :–95 °C
- :0,79 g/cm³
- :–20 °C (closed cup)
- :Stue temperatur
- :200-662-2
- :1090
- :3,II
- :01-2119471330-49
Documents
ACRO40010
SDS
- Appearance:Clear liquid
- Water:≤0.5 % (K.F.)
- Solubility:(in water) Passes test
- GC:≥99.5 %
- UV:at 330 nm A: ≤1
- Color scale:≤10 APHA
- Residue after evaporation:≤0.001 %
- Isopropyl alcohol:≤0.05 %
- Titrable acid:≤0.0003 meq/g
- Titrable base:≤0.0006 meq/g
- Aldehyde (HCHO):≤0.002 %
- Methanol:≤0.05 %
- KMnO4-reducing substance:Passes test
- Pakning:Glass bottle
- Pakn:500 mL
- Cat. No.:ACRO400105000
- Miljøcertificering:Chem21 Solvent Selection Guide recommended after discussion
- Environmentally Preferable:Sikrere sundhed for mennesker og miljø
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



