PTFE balls, BOLA
Balls, PTFE, 20 mm
BOHLH964-18
Made from solid PTFE, with smooth surface.
For extension of surfaces of e.g. distillation apparatus; as splash guard.
- Ø:20 mm
- Beskrivelse:Balls
- Materiale:PTFE
- Seq:010