Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Stirrer bearings, PTFE

Stirrer bearings, PTFE

Omrørerleje med pakningsring, 45/40 NS, GL 32, PTFE, 16 mm skaft, 45/40, 86 mm
Catalog # BOHLC424-18
Supplier:   Bohlender
Stirrer bearings, PTFE
Catalog # BOHLC424-18
Supplier:  Bohlender
Stirrer bearings, PTFE
Stirrer bearings, PTFE

Omrørerleje med pakningsring, 45/40 NS, GL 32, PTFE, 16 mm skaft, 45/40, 86 mm

Price
802,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents

These bearings have sealing rings to ensure a perfect seal. A special gasket made of PTFE and a FPM o-ring which is compressed by a GL screw cap ensure the stirrer shaft is sealed.

  • Cone can be removed easily from the socket
  • Ground joint does not stick and danger of breaking is reduced
  • Gasket can be exchanged when worn
Specifications
  • Til:
    16 mm skaft
  • Beskrivelse:
    Omrørerleje med pakningsring, 45/40 NS, GL 32, PTFE
  • Længde:
    86 mm
  • Seq:
    009
  • NS prop:
    45/40