Orders LinkContactUsLinkComponent
Powder funnels
200 mm, 180 mm, 30 mm
Katalog # 221-1619
Leverandør:   BOCHEM
Powder funnels
Katalog # 221-1619
Leverandør:  BOCHEM
Powder funnels
Powder funnels
200 mm, 180 mm, 30 mm
Price
1.520,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel 18/10.
  • Smooth internal walls
  • Robust and strong
  • Good chemical resistance, extremely high thermal resistance
With short, wide stem.
Specifikationer
  • Tragt udv. Ø:
    200 mm
  • Længde:
    180 mm
  • Stilk udv. Ø:
    30 mm