Orders LinkContactUsLinkComponent
Measuring cylinders, conical
Measuring cylinders, conical
2000 ml, 185 mm, 160 mm
Katalog # BOCH8663
Leverandør:   BOCHEM
Measuring cylinders, conical
Katalog # BOCH8663
Leverandør:  BOCHEM
Measuring cylinders, conical
Measuring cylinders, conical
2000 ml, 185 mm, 160 mm
Price
559,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel 18/10.
  • With base and handle
  • Conical, graduated
Specifikationer
  • Ø:
    160 mm
  • Kapacitet:
    2000 ml
  • Højde:
    185 mm