Orders LinkContactUsLinkComponent
Measuring cylinders, conical
Measuring cylinders, conical
1500 ml, 160 mm, 150 mm
Katalog # BOCH8662
Leverandør:   BOCHEM
Measuring cylinders, conical
Katalog # BOCH8662
Leverandør:  BOCHEM
Measuring cylinders, conical
Measuring cylinders, conical
1500 ml, 160 mm, 150 mm
Price
455,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Stainless steel 18/10.
  • With base and handle
  • Conical, graduated
Specifikationer
  • Ø:
    150 mm
  • Kapacitet:
    1500 ml
  • Højde:
    160 mm