Orders LinkContactUsLinkComponent
Evaporating dishes, flat shape
Evaporating dishes, flat shape
75 ml, 20 mm, 70 mm
Catalog # BOCH8571
Supplier:   BOCHEM
Evaporating dishes, flat shape
Catalog # BOCH8571
Supplier:  BOCHEM
Evaporating dishes, flat shape
Evaporating dishes, flat shape
75 ml, 20 mm, 70 mm
Price
227,00 kr
Log in for availability
1 * 1 Items
Product Details & Documents
Evaporating dishes are made of stainless steel 18/10.
  • Very good chemical resistance
  • Very good temperature resistance
  • Easy to clean
Specifications
  • Udv.Ø:
    70 mm
  • Kapacitet:
    75 ml
  • Højde:
    20 mm