Instrument trays
300×200×15 mm
Katalog # BOCH8484
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Made of 18/10 steel.
- Very good chemical resistance
- Very good temperature resistance
- Sturdy
Documents
Certifikater
Søg efter partinummerSpecifikationer
- B×D×H:300×200×15 mm