Spande
Låg til spande med inddeling 12 l, rustfrit stål 18/10, Ø 300 mm
Katalog # 216-0232
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Buckets, stainless steel 18/10.
- Very good chemical resistance, very good thermal resistance
- High quality, very stable
- Matching lids available optionally
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.
Specifikationer
- Beskrivelse:Låg til spande med inddeling 12 l, rustfrit stål 18/10, Ø 300 mm
- Seq:10