Orders LinkContactUsLinkComponent
Spande
15 L, 310 mm, 305 mm
Katalog # 216-1602
Leverandør:   BOCHEM
Spande
Katalog # 216-1602
Leverandør:  BOCHEM
Spande
Spande
15 L, 310 mm, 305 mm
Price
1.820,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Buckets, stainless steel 18/10.
  • Very good chemical resistance, very good thermal resistance
  • High quality, very stable
  • Matching lids available optionally
For use in laboratories and industry. Graduated with handle. Seamless handle tabs and smooth surfaces for fast, easy cleaning.
Specifikationer
  • Kapacitet:
    15 L
  • Højde:
    305 mm
  • Seq:
    03
  • Ø udv.:
    310 mm