Orders LinkContactUsLinkComponent
Retort rings, open
Open retort ring, 220 mm length, 100 mm
Catalog # BOCH5512
Supplier:   BOCHEM
Retort rings, open
Catalog # BOCH5512
Supplier:  BOCHEM
Retort rings, open
Retort rings, open
Open retort ring, 220 mm length, 100 mm
Price
kr226.00
Log in for availability
1 * 1 Items
Product Details & Documents
Retort rings made from stainless steel 18/10.
Specifications
  • Ø indv.:
    100 mm
  • Beskrivelse:
    Open retort ring, 220 mm length
  • Seq:
    03