Orders LinkContactUsLinkComponent
Retort rings, open
Retort rings, open
Open retort ring, 160 mm length, 50 mm
Katalog # BOCH5510
Leverandør:   BOCHEM
Retort rings, open
Katalog # BOCH5510
Leverandør:  BOCHEM
Retort rings, open
Retort rings, open
Open retort ring, 160 mm length, 50 mm
Price
198,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Retort rings made from stainless steel 18/10.
Specifikationer
  • Ø indv.:
    50 mm
  • Beskrivelse:
    Open retort ring, 160 mm length
  • Seq:
    01