Spoon spatulas
210 mm, 25×12 mm
Catalog # 231-2227
Supplier: BOCHEM
Jump to:
Product Details & Documents
PTFE coated double spatulas with one spoon end and one flat end.
Specifications
- Længde:210 mm
- Seq:02
- Skestr.:25×12 mm
PTFE coated double spatulas with one spoon end and one flat end.