Vejeskeer
Vejeske med greb, 200 mm, 31 mm
Catalog # BOCH3600
Supplier: BOCHEM
Jump to:
Product Details & Documents
These weighing scoops with knob are made made of 18/10 stainless steel. Available with or without green PTFE coating.
Specifications
- Bredde:31 mm
- Beskrivelse:Vejeske med greb
- Længde:200 mm
- Seq:01