Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Apotekerspatel

Apotekerspatel

365 mm, 250 mm, 38 mm
Catalog # 231-1077
Supplier:   BOCHEM
Apotekerspatel
Catalog # 231-1077
Supplier:  BOCHEM
Apotekerspatel
Apotekerspatel

365 mm, 250 mm, 38 mm

Price
84,50 kr
Log in for availability
1 * 1 Items
Product Details & Documents

Rustfrit stål blad med træhåndtag.

  • Blad med god varmeresistens
  • God kemisk resistens
  • Rengøres nemt og hurtigt

Documents

Specifications
  • Længde:
    365 mm
  • Bladbredde:
    38 mm
  • Klinge længde:
    250 mm
Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.