Forceps for cover glasses
Pincet til dækglas, bøjet, rustfrit stål, 105 mm
Katalog # 232-1262
Leverandør: BOCHEM
Gå til:
Product Details & Documents
Stainless steel or stainless steel with PTFE coating.
- For easy, secure handling of cover glasses
- With 6 mm wide tips
- Can also be used for other laboratory equipment, such as filter discs
- PTFE coated design with particularly smooth surface to prevent contamination and enable simple cleaning
Specifikationer
- Beskrivelse:Pincet til dækglas, bøjet, rustfrit stål
- Længde:105 mm
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



