Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
 

Bromphenol rød, pure

Glass bottle for solids, 5 g
Katalog # ACRO304550050
Leverandør:   Thermo Fisher Scientific
Bromphenol rød, pure
Katalog # ACRO304550050
CAS nummer:  2800-80-8
Bromphenol rød, pure
Bromphenol rød, pure

Glass bottle for solids, 5 g

CAS nummer:  2800-80-8
Price
669,00 kr
Log ind for tilgængelighed
1 * 5 g
Product Details & Documents
Synonymer:  4,4'-(3H-2,1-Benzoxathiol-3-ylidene)bis[2-bromophenol] S,S-dioxide
  • CAS nummer:2800-80-8
  • Formel:C₁₉H₁₂Br₂O₅S
  • MDL nummer:MFCD00023014
  • Molecular Weight:512.18 g/mol
  • Smeltende Pt:230 °C
  • EINECS:220-538-1

Documents

Specifikationer

Specifikationstestresultater

  • Appearance:
    Red-brown to brown Powder
  • Infrared spectrum:
    Conforms
  • Loss on drying:
    ≤5 % (110°C)
  • Solubility:
    (1 % in ethanol) Clear
  • Visual transition interval:
    from pH 5.2 (yellow-red) to pH 7.0 (purple)

Specifikationer

  • Pakning:
    Glass bottle for solids
  • Pakn:
    5 g

Specifikationer

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.