3-Fluor-4-nitrophenol 99%
Glass bottle for solids, 5 g
Leverandør: Thermo Fisher Scientific
3-Fluor-4-nitrophenol 99%
Katalog # ACRO303370050
Leverandør: Thermo Fisher Scientific
CAS nummer: 394-41-2
Gå til:
Product Details & Documents
- CAS nummer:394-41-2
- Formel:C₆H₄FNO₃
- MDL nummer:MFCD00041251
- Molecular Weight:157.1 g/mol
- Smeltende Pt:92…95 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:206-895-6
- FN:2811
Documents
ACRO30337
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Light yellow to brown Powder
- Infrared spectrum:Conforms
- Melting point:91 to 95 °C
- GC:≥98.5 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:5 g
Specifikationer
Recommendations
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



