1-Octen 99+%
Glass bottle, 100 mL
Leverandør: Thermo Fisher Scientific
Gå til:
Product Details & Documents
- CAS nummer:111-66-0
- Formel:C₈H₁₆
- MDL nummer:MFCD00009548
- Molecular Weight:112.22 g/mol
- Merck Index:13,01770
- Kogende Pt:121…123 °C (760 mmHg)
- Smeltende Pt:–102 °C
- Tæthed:0,715 g/cm³
- Flash Pt:13 °C
- Opbevaringstemperatur:Stue temperatur
- EINECS:203-893-7
- FN:3295
- ADR:3,II
Documents
ACRO30125
SDS
Specifikationer
Specifikationstestresultater
- Appearance:Clear liquid
- Infrared spectrum:Conforms
- Refractive index:1.4080 to 1.4100 (20°C, 589 nm)
- Water:≤0.005 %
- GC:≥99.0 %
Specifikationer
- Pakning:Glass bottle
- Pakn:100 mL
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



