Orders LinkContactUsLinkComponent
Adapters, receiver, plain bent, Quickfit®
Adapters, receiver, plain bent, Quickfit®
19/26 NS, 19/26 NS
Katalog # 271-0015
Leverandør:   SciLabware
Adapters, receiver, plain bent, Quickfit®
Katalog # 271-0015
Leverandør:  SciLabware
Adapters, receiver, plain bent, Quickfit®
Adapters, receiver, plain bent, Quickfit®
19/26 NS, 19/26 NS
Price
376,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
Receiver, delivery, PYREX® borosilicate glass
  • Excellent chemical resistance
  • High temperature resistance
  • Minimal thermal expansion, giving relatively high resistance to temperature changes
Plain bend without vent.
Specifikationer
  • Konusstr.:
    19/26 NS
  • Kappestr.:
    19/26 NS
  • Seq:
    02