Orders LinkContactUsLinkComponent
Watch glasses
Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 125 mm
Katalog # 216-2752
Leverandør:   SciLabware
Watch glasses
Katalog # 216-2752
Leverandør:  SciLabware
Watch glasses
Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 125 mm
Price
209,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
  • Meget god kemisk resistens
  • Høj temperaturresistens
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifikationer
  • Ø:
    125 mm
  • Beskrivelse:
    Watch glass, PYREX® borosilicate glass with firepolished edges
  • Seq:
    08