Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 50 mm
Catalog # 216-2748
Supplier: SciLabware
Jump to:
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
- Meget god kemisk resistens
- Høj temperaturresistens
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifications
- Ø:50 mm
- Beskrivelse:Watch glass, PYREX® borosilicate glass with firepolished edges
- Seq:02