Watch glasses
Watch glass, PYREX® borosilicate glass with firepolished edges, 40 mm
Katalog # 216-2770
Leverandør: SciLabware
Gå til:
Product Details & Documents
PYREX®, borosilicate glass, with fused edges.
- Meget god kemisk resistens
- Høj temperaturresistens
For evaporation work and covering heated beakers. Fire-polished edges to reduce chipping and breakage.
Specifikationer
- Ø:40 mm
- Beskrivelse:Watch glass, PYREX® borosilicate glass with firepolished edges
- Seq:01