Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Cyclohexan 99.8% for HPLC

Cyclohexan 99.8% for HPLC

Glass bottle, 2,5 L
Catalog # ACRO279590025
Irritant
Health
Environment
Flammable
Cyclohexan 99.8% for HPLC
Catalog # ACRO279590025
CAS Number:  110-82-7
Cyclohexan 99.8% for HPLC
Cyclohexan 99.8% for HPLC

Glass bottle, 2,5 L

CAS Number:  110-82-7
Price
2.150,00 kr
Log in for availability
1 * 2,5 L
Product Details & Documents
Synonyms:  Hexahydrobenzene, Hexamethylene
  • CAS Number:110-82-7
  • Formula:C₆H₁₂
  • MDL Number:MFCD00003814
  • Molecular Weight:84.16 g/mol
  • Merck Index:13,02752
  • Boiling Pt:81 °C (1013 hPa)
  • Melting Pt:6,5 °C
  • Density:0,77 g/cm³
  • Flash Pt:–18 °C
  • Storage Temperature:Stue temperatur
  • EINECS:203-806-2
  • UN:1145
  • ADR:3,II

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.01 %
  • GC:
    ≥99.8 %
  • UV:
    at 200 nm A: ≤0.3
  • Color scale:
    ≤10 APHA
  • Residue after evaporation:
    ≤0.0002 %
  • Acidity (CH3COOH):
    ≤0.0002 meq/g

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    2,5 L

Specifications

Recommendations

Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".

Otherwise, you will receive generic recommendations.