Cyclohexan 99.8% for HPLC
Glass bottle, 1 L
: Hexahydrobenzene, Hexamethylene
- :110-82-7
- :C₆H₁₂
- :MFCD00003814
- :84.16 g/mol
- :13,02752
- :81 °C (1013 hPa)
- :6,5 °C
- :0,77 g/cm³
- :–18 °C
- :Stue temperatur
- :203-806-2
- :1145
- :3,II
- Appearance:Clear liquid
- Water:≤0.01 %
- GC:≥99.8 %
- UV:at 200 nm A: ≤0.3
- Color scale:≤10 APHA
- Residue after evaporation:≤0.0002 %
- Acidity (CH3COOH):≤0.0002 meq/g
- Pakning:Glass bottle
- Pakn:1 L



