Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Weighing boats, PYREX®

Weighing boats, PYREX®

6 ml, 85 mm
Katalog # 611-0010
Leverandør:   SciLabware
Weighing boats, PYREX®
Katalog # 611-0010
Leverandør:  SciLabware
Weighing boats, PYREX®
Weighing boats, PYREX®

6 ml, 85 mm

Price
280,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents

Borosilicate glass.

  • Ideal for weighing small quantities of powder
  • With hollow stem to facilitate introduction of the powder into narrow neck vessels e.g. volumetric flasks
  • Flat base ensures stability on the balance pan
Specifikationer
  • Kapacitet:
    6 ml
  • Længde:
    85 mm
  • Seq:
    03