Orders LinkContactUsLinkComponent
Jacketed beakers, tall form
Jacketed beakers, tall form
1000 ml, 235 mm, 110 mm
Katalog # 213-0042
Leverandør:   SciLabware
Jacketed beakers, tall form
Katalog # 213-0042
Leverandør:  SciLabware
Jacketed beakers, tall form
Jacketed beakers, tall form
1000 ml, 235 mm, 110 mm
Price
4.510,00 kr
Log ind for tilgængelighed
1 * 1 Items
Product Details & Documents
PYREX®, borosilicate glass, without spout.
  • Very good chemical resistance
  • Very good thermal resistance
  • With screw thread and plastic threaded tubing connectors
  • Tube nozzles with 11 mm ext. diameter (Ø) for 250 and 600 ml beakers and 14 mm ext. diameter (Ø) for 1000 and 2000 ml beakers
For maintaining liquid samples at a constant temperature. The temperature controlling medium is pumped through the double-walled jacket by means of a recirculating thermostat.
Documents
Specifikationer
  • Udv.Ø:
    110 mm
  • Kapacitet:
    1000 ml
  • Højde:
    235 mm
  • Seq:
    03