3'-Chlorpropiophenon 98%
Glass bottle for solids, 25 g
Leverandør: Thermo Fisher Scientific
3'-Chlorpropiophenon 98%
Katalog # ACRO276580250
Leverandør: Thermo Fisher Scientific
CAS nummer: 34841-35-5
Gå til:
Product Details & Documents
Synonymer: m-Chloropropiophenone, 1-(3-Chlorophenyl)propan-1-one
- CAS nummer:34841-35-5
- Formel:C₉H₉ClO
- MDL nummer:MFCD00009925
- Molecular Weight:168.62 g/mol
- Kogende Pt:124 °C (14 mmHg)
- Smeltende Pt:43…47 °C
- Flash Pt:>110 °C
- EINECS:252-242-3
Documents
ACRO27658
SDS
Specifikationer
Specifikationstestresultater
- Appearance:White to light yellow Crystalline solid
- Infrared spectrum:Conforms
- Melting point:43 to 47 °C
- GC:≥97.5 %
Specifikationer
- Pakning:Glass bottle for solids
- Pakn:25 g
Specifikationer
Anbefalinger
Recommendations will be personalized based on your shopping preferences only if you have given your consent by enabling "Enhance my Shopping Experience" on the "Personal Info page".
Otherwise, you will receive generic recommendations.



