Hydroxylammoniumchlorid 99+%
Plastic bottle, 100 g
Leverandør: Thermo Fisher Scientific
Hydroxylammoniumchlorid 99+%
Katalog # ACRO270101000
Leverandør: Thermo Fisher Scientific
CAS nummer: 5470-11-1
Gå til:
Product Details & Documents
Synonymer: Hydroxylamine HCl, Hydroxylamine hydrochloride
- CAS nummer:5470-11-1
- Formel:NH₂OH·HCl
- MDL nummer:MFCD00051089
- Molecular Weight:69.49 g/mol
- Merck Index:13,04853
- Kogende Pt:306 °C (1013 hPa)
- Smeltende Pt:155…158 °C (dec.)
- Tæthed:1,67 g/cm³ (20 °C)
- Opbevaringstemperatur:Stue temperatur
- EINECS:226-798-2
- FN:2923
- ADR:8,III
Documents
Specifikationer
Specifikationstestresultater
- Appearance:White Crystalline powder or crystals and/or chunks
- Infrared spectrum:Conforms
- Titration with NaOH:99.0 to 101.0 %
- Sulfate (SO4):≤0.005 %
- Iron (Fe):≤0.001 %
Specifikationer
- Pakning:Plastic bottle
- Pakn:100 g