Ordreindtastning
Denmark
Orders LinkContactUsLinkComponent
Dichlormethan 99.8% stabilised for HPLC

Dichlormethan 99.8% stabilised for HPLC

Glass bottle, 1 L
Catalog # ACRO268330010
Irritant
Health
Dichlormethan 99.8% stabilised for HPLC
Catalog # ACRO268330010
CAS Number:  75-09-2
Dichlormethan 99.8% stabilised for HPLC
Dichlormethan 99.8% stabilised for HPLC

Glass bottle, 1 L

CAS Number:  75-09-2
Price
kr719.00
Log in for availability
1 * 1 L
Product Details & Documents
Synonyms:  Methylenchlorid
  • CAS Number:75-09-2
  • Formula:CH₂Cl₂
  • MDL Number:MFCD00000881
  • Molecular Weight:84.93 g/mol
  • Merck Index:12,06140
  • Boiling Pt:39,8 °C (1013 hPa)
  • Melting Pt:–97 °C
  • Density:1,325 g/cm³
  • Storage Temperature:Stue temperatur
  • EINECS:200-838-9
  • ADR:6.1,III
  •  :stabiliseret med amylen
  • UN:1593

Documents

Specifications

Specification Test Results

  • Appearance:
    Clear liquid
  • Water:
    ≤0.02 %
  • GC:
    ≥99.8 %
  • UV:
    at 233 nm A: ≤1
  • Color scale:
    ≤10 APHA
  • Stabilizer:
    About 100 ppm Amylene
  • Residue after evaporation:
    ≤0.0002 %
  • Acidity:
    ≤0.0001 meq/g
  • Filtered to 0.2 micrometer:
    Filtered
  • Free chlorine:
    ≤0.0005 %

Specifications

  • Pakning:
    Glass bottle
  • Pakn:
    1 L

Specifications